Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetracaine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. T168825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T168825G 25 g
T16885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T168825G Supplier TCI America Supplier No. T168825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 136-47-0
Molecular Formula C15H25ClN2O2
Molecular Weight (g/mol) 300.827
MDL Number MFCD00038912
InChI Key PPWHTZKZQNXVAE-UHFFFAOYSA-N
Synonym tetracaine hydrochloride, tetracaine hcl, butethanol, butylocaine, anethain, anethaine, curtacain, decicain, decicaine, dessicain
PubChem CID 8695
IUPAC Name 2-(dimethylamino)ethyl 4-(butylamino)benzoate;hydrochloride
SMILES CCCCNC1=CC=C(C=C1)C(=O)OCCN(C)C.Cl

Specifications

Melting Point 149°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 300.83
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetracaine Hydrochloride
RTECSNumber DG4900000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.