Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetracaine Related Compound C, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023023 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-102-3023 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-3023 Supplier MilliporeSigma Supelco Supplier No. PHR2261100MG
Only null left
Add to Cart
Add to Cart

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

TRUSTED_SUSTAINABILITY

Specifications

CAS 71839-12-8
Flash Point Not Applicable
Molecular Formula C12H17NO2
Linear Formula C12H17NO2
Quantity 100 mg
Synonym Methyl 4-(butylamino)benzoate; 4-(Butylamino)benzoic acid methyl ester
Formula Weight 207.27
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.