Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich TETRACALCIUM PHOSPHATE (TTCP) POWDER ≥98% <40 μm PARTICLE SIZE

Catalog No. 91885710G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-885-710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-885-710G Supplier Sigma Aldrich Supplier No. 91885710G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetracalcium phosphate (TTCP) is used in calcium hydroxide cements, and hydrolyzes slowly to form calcium hydroxide and a thin insoluble apatite layer that prevents further reaction. TTCP has a calcium phosphate phase with a Ca/P ratio greater than stoichiometric hydroxyapatite (HA) which makes it useful as a ceramic biomaterial.

Specifications

CAS 1306-01-0
Linear Formula Ca4(PO4)2 O
Quantity 10 g
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.