Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachloro-o-benzoquinone, 97%, Thermo Scientific™

Catalog No. AC137991000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC137991000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC137991000 Supplier Thermo Scientific Chemicals Supplier No. 137991000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2435-53-2
Molecular Formula C6Cl4O2
Molecular Weight (g/mol) 245.86
MDL Number MFCD00001646
InChI Key VRGCYEIGVVTZCC-UHFFFAOYSA-N
Synonym o-chloranil, tetrachloro-o-benzoquinone, tetrachloro-o-quinone, 2-chloranil, tetrachloro-1,2-benzoquinone, o-chloroanil, 3,4,5,6-tetrachloro-1,2-benzoquinone, isochloranil, 3,5-cyclohexadiene-1,2-dione, 3,4,5,6-tetrachloro, ortho-chloranil
PubChem CID 73252
IUPAC Name tetrachlorocyclohexa-3,5-diene-1,2-dione
SMILES ClC1=C(Cl)C(=O)C(=O)C(Cl)=C1Cl

Specifications

Melting Point 125.0°C to 129.0°C
Linear Formula C6Cl4(=O)2
Quantity 100 g
Solubility Information Solubility in water: insoluble. Other solubilities: 10g/L chloroform (20°C)
Formula Weight 245.88
Chemical Name or Material Tetrachloro-o-benzoquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.