Learn More
Medchemexpress LLC Tetrachloroauric acid | 16903-35-8 | MFCD00011322 | 97.0% | 339.79 g/mol | HAuCl4 | 100 MG
SDP
Catalog No. 5000299440
Encompass_Preferred
Tetrachloroauric acid (HAuCl4, CAS 16903-35-8) is a gold(III) inorganic salt used as a precursor in gold chemistry. Supplied as a high-purity reagent, it is commonly used for gold plating, synthesis of gold nanoparticles, and preparation of gold complexes. Handle using appropriate laboratory safety procedures and consult the SDS for hazards and storage.
  • Typical purity 97.0%.
  • Molecular formula HAuCl4; molecular weight 339.79 g/mol.
  • Used as a precursor for gold plating and gold nanoparticle synthesis.
  • Available in 100 mg, 250 mg, 500 mg, and 1 g pack sizes.
  • Suitable for research and laboratory use; follow SDS for handling and storage.
Catalog No. 50-002-99440 Supplier Medchemexpress LLC Supplier No. HYW035091100MG
Add to Cart
Add to Cart
Tetrachloroauric acid (HAuCl4, CAS 16903-35-8) is a gold(III) inorganic salt used as a precursor in gold chemistry. Supplied as a high-purity reagent, it is commonly used for gold plating, synthesis of gold nanoparticles, and preparation of gold complexes. Handle using appropriate laboratory safety procedures and consult the SDS for hazards and storage.
  • Typical purity 97.0%.
  • Molecular formula HAuCl4; molecular weight 339.79 g/mol.
  • Used as a precursor for gold plating and gold nanoparticle synthesis.
  • Available in 100 mg, 250 mg, 500 mg, and 1 g pack sizes.
  • Suitable for research and laboratory use; follow SDS for handling and storage.