Learn More
Medchemexpress LLC Tetrachloroauric acid | 16903-35-8 | MFCD00011322 | 97.0% | 339.79 g·mol⁻¹ | HAuCl4 | 250 MG
SDP
Catalog No. 5000303501
Encompass_Preferred
Tetrachloroauric acid (HAuCl4) is an inorganic gold(III) reagent commonly supplied as a hydrate. It serves as a source of Au(III) for materials chemistry, including gold plating and the synthesis of gold nanoparticles. The reagent is provided at high purity for laboratory research and materials applications.
  • High purity reagent (97%).
  • Suitable for gold plating and nanoparticle synthesis.
  • Molecular weight 339.79 g·mol⁻¹ for stoichiometric calculations.
  • Hydrate form with good water solubility.
  • Small pack size optimized for research use.
Catalog No. 50-003-03501 Supplier Medchemexpress LLC Supplier No. 250MGHYW035091
Add to Cart
Add to Cart
Tetrachloroauric acid (HAuCl4) is an inorganic gold(III) reagent commonly supplied as a hydrate. It serves as a source of Au(III) for materials chemistry, including gold plating and the synthesis of gold nanoparticles. The reagent is provided at high purity for laboratory research and materials applications.
  • High purity reagent (97%).
  • Suitable for gold plating and nanoparticle synthesis.
  • Molecular weight 339.79 g·mol⁻¹ for stoichiometric calculations.
  • Hydrate form with good water solubility.
  • Small pack size optimized for research use.