Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachlorobis(tetrahydrofuran)zirconium(IV), Thermo Scientific Chemicals

Catalog No. AA3955004
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3955004 2 g
AA3955009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3955004 Supplier Thermo Scientific Chemicals Supplier No. 03955004
Add to Cart

missing translation for 'validMainframeMsg'

It is an important organic intermediate and can be used in agrochemicals, paharmaceuticals and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21959-01-3
Molecular Formula C8H16Cl4O2Zr
Molecular Weight (g/mol) 377.238
MDL Number MFCD00145367
InChI Key VDJJKYYENYIHFF-UHFFFAOYSA-J
Synonym tetrachlorobis tetrahydrofuran zirconium, zrcl4 thf 2, oxolane; tetrachlorozirconium, tetrachlorobis tetrahydrofurane zirconium, zirconium tetrachloride tetrahydrofuran complex, zirconium iv chloride tetrahydrofuran complex, bis tetrahydrofuran ; zirconium iv chloride, zirconium iv chloride tetrahydrofuran complex 1:2
PubChem CID 2734040
IUPAC Name oxolane;tetrachlorozirconium
SMILES C1CCOC1.C1CCOC1.Cl[Zr](Cl)(Cl)Cl

Specifications

Melting Point 175°C to 177°C
Color White
Quantity 2 g
Physical Form Crystalline
UN Number UN2925
Solubility Information Reacts with water.
Formula Weight 377.25
Chemical Name or Material Tetrachlorobis(tetrahydrofuran)zirconium(IV)
Hazard Category H302+H312+H332-H314
Hazard Statement GHS H Statement
H228-H314-H318
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.