Learn More
Medchemexpress LLC HY-W006000 500mg Medchemexpress, Tetrachlorocatechol CAS:1198-55-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW006000500MG
Encompass_Preferred
Medchemexpress, HY-W006000 500mg Tetrachlorocatechol CAS:1198-55-6 Tetrachlorocatechol is a metabolite of pentachlorophenol. Tetrachlorocatechol is one of the most toxic chlorinated catechol produced by the chlorobleaching of pulp and frequently found in the kraft pulp mill effluents. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3230