Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachlorocyclopropene, 98%

Catalog No. AC420190250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC420190250 25 g Glass bottle
AC420190050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC420190250 Supplier Thermo Scientific Chemicals Supplier No. 420190250
  • $780.00
    $663.65 / Each of 1

    Save $116.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6262-42-6 | C3Cl4 | 177.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6262-42-6
Molecular Formula C3Cl4
Molecular Weight (g/mol) 177.84
MDL Number MFCD00001267
InChI Key BLZOHTXDDOAASQ-UHFFFAOYSA-N
Synonym tetrachlorocyclopropene, cyclopropene, tetrachloro, 1,2,3,3-tetrachloro-cyclopropene, tetrachlorocycloprop-1-ene, acmc-20ajjz, dsstox_cid_1660, dsstox_rid_76272, dsstox_gsid_21660, cyclopropene,1,2,3,3-tetrachloro
PubChem CID 80428
IUPAC Name 1,2,3,3-tetrachlorocyclopropene
SMILES C1(=C(C1(Cl)Cl)Cl)Cl

Specifications

Color Colorless
Boiling Point 125°C to 130°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5063 to 1.5083
Quantity 25 g
Formula Weight 177.84
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tetrachlorocyclopropene, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 228-418-0

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.