Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachloroethylene, 99%

Catalog No. p-7024851
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB20089AE 100 mL
AAB20089AP 500 mL
AAB200890F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB20089AE Supplier Thermo Scientific Chemicals Supplier No. B20089AE
Only null left
Add to Cart

CAS: 127-18-4 | C2Cl4 | 165.82 g/mol

Tetrachloroethylene is used as a solvent. It is involved in dry cleaning as well as in degreasing of metal parts. It finds application as a neutrino detector. It is utilized as a paint stripper and as a spot remover in consumer products. It acts as a precursor for the manufacture of other chemicals. It plays an important role in water repellants, paint removers, printing inks, glues, sealants, polishes and lubricants. As a film-forming electrolyte additive, it is used in the manufacture of lithium ion batteries and is actively involved as an extraction solvent in the determination of oil and grease in water by FT-IR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrachloroethylene is used as a solvent. It is involved in dry cleaning as well as in degreasing of metal parts. It finds application as a neutrino detector. It is utilized as a paint stripper and as a spot remover in consumer products. It acts as a precursor for the manufacture of other chemicals. It plays an important role in water repellants, paint removers, printing inks, glues, sealants, polishes and lubricants. As a film-forming electrolyte additive, it is used in the manufacture of lithium ion batteries and is actively involved as an extraction solvent in the determination of oil and grease in water by FT-IR.

Solubility
Miscible with alcohol, ether, chloroform, benzene and hexane. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases, alkali metals and aluminum.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-18-4
Molecular Formula C2Cl4
Molecular Weight (g/mol) 165.82
MDL Number MFCD00000834
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
Synonym tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
PubChem CID 31373
ChEBI CHEBI:17300
SMILES ClC(Cl)=C(Cl)Cl

Specifications

Melting Point -22°C
Density 1.623
Boiling Point 120°C to 121°C
Odor Ether-like
Assay Percent Range 0.99
Refractive Index 1.5055
Linear Formula Cl2C=CCl2
Quantity 100 mL
UN Number UN1897
Beilstein 1361721
Merck Index 14,9190
Solubility Information Miscible with alcohol,ether,chloroform,benzene and hexane. Slightly miscible with water.
IUPAC Name tetrachloroethene
Formula Weight 165.83
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tetrachloroethylene
Show More Show Less
Hazard Category H315-H317-H319-H336-H350-H373
Hazard Statement GHS H Statement
H351
Suspected of causing cancer.
Precautionary Statement P201-P202-P260-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TETRACHLOROETHYLENE
EINECSNumber 204-825-9
RTECSNumber KX3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.