Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachloroethylene, 99%, extra pure, ACROS Organics™

Click to view available options
Quantity:
500mL
1L
2.5L
10L
Packaging:
Glass Bottle
Plastic drum
127-18-4
C2Cl4
165.83
MFCD00000834
CYTYCFOTNPOANT-UHFFFAOYSA-N
tetrachloroethylene,tetrachloroethene,perchloroethylene,perchlorethylene,perc,ethene, tetrachloro,tetrachlorethylene,ethylene tetrachloride,carbon dichloride,ankilostin
31373
CHEBI:17300
1,1,2,2-tetrachloroethene
C(=C(Cl)Cl)(Cl)Cl
127-18-4
C2Cl4
165.83
MFCD00000834
CYTYCFOTNPOANT-UHFFFAOYSA-N
tetrachloroethylene,tetrachloroethene,perchloroethylene,perchlorethylene,perc,ethene, tetrachloro,tetrachlorethylene,ethylene tetrachloride,carbon dichloride,ankilostin
31373
CHEBI:17300
1,1,2,2-tetrachloroethene
C(=C(Cl)Cl)(Cl)Cl
Viscosity 0.89 mPa.s (20°C)
Boiling Point 120.0°C to 122.0°C
Specific Gravity 1.625
Residue after Evaporation 0.005% max.
Refractive Index 1.505
Water 0.01% max.
Quantity 2.5L
Packaging Glass Bottle
Alkalinity 0.003% max. (NaOH)
Density 1.6250g/mL
Color Undesignated
Melting Point -22.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula Cl2C=CCl2
Beilstein 01, 187
Merck Index 15, 9335
Solubility Information Solubility in water: 0.15g/L (20°C). Other solubilities: miscible with alcohol, ether, oils, chloroform, and benzene
Formula Weight 165.83
Physical Form Liquid
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Tetrachloroethylene
Show More Show Less

GHS P Statement
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause an allergic skin reaction.
May cause drowsiness or dizziness.

GHS P Statement
Use personal protective equipment as required.
Avoid release to the environment.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning