Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachloroethylene, CHROMASOLV™, for HPLC, ≥99.9%, Solstice
GSA_VA

Catalog No. 6001662 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-016-62 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-016-62 Supplier Solstice Supplier No. 2703931L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™, for HPLC,≥99.9%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-18-4
Molecular Formula C2Cl4
Molecular Weight (g/mol) 165.82
MDL Number MFCD00000834
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
Synonym tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
PubChem CID 31373
ChEBI CHEBI:17300
SMILES ClC(Cl)=C(Cl)Cl

Specifications

Absorbance max. 1.0 at 290 nm, max. 0.03 at 400 nm, max. 0.05 at 350 nm, max. 0.10 at 305 nm, max. 0.20 at 300 nm, max. 0.30 at 295 nm
CAS Max % 100.0000%
Melting Point -22°C (literature)
Density 1.623 g/mL at 25 °C(lit.)
Boiling Point 120°C to 122°C
Packaging Glass Bottle
Linear Formula CCl2=CCl2
Quantity 1 L
UN Number UN1897
Beilstein 1361721
IUPAC Name 1,1,2,2-tetrachloroethene
Formula Weight 165.83
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Tetrachloroethylene, CHROMASOLV™, for HPLC, ≥99.9%
Show More Show Less
ShelfLife 900 days from date of manufacture
DOTInformation Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Tetrachloroethylene
EINECSNumber 204-825-9
Recommended Storage Room Temp
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.