Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachloroethylene (Technical), Fisher Chemical™

Catalog No. C18220 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
20 L
Packaging:
Lined Steel Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C182-20 20 L Lined Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C182-20

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 127-18-4
Molecular Formula C2Cl4
Molecular Weight (g/mol) 165.82
MDL Number MFCD00000834
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
Synonym tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
PubChem CID 31373
ChEBI CHEBI:17300
IUPAC Name tetrachloroethene
SMILES ClC(Cl)=C(Cl)Cl

Specifications

Decomposition Information >150°C
Melting Point -22°C
Color Colorless
Density 1.625g/cm³
Boiling Point 120°C
Packaging Lined Steel Drum
Linear Formula CCl2:CCl2
Quantity 20 L
Vapor Pressure 18mbar at 20°C
Formula Weight 165.83
Grade Technical
Physical Form Liquid
Chemical Name or Material Tetrachloroethylene
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Suspect cancer hazard. Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:0
Instability:0
DOTInformation DOT Class 6.1, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.