Learn More
Cayman Chemical Tetrachlorohydroquinone, Purity: ≥98%, CAS: 87-87-6, 10 g
SDP
Catalog No. 502993833
Encompass_Preferred
A metabolite of pentachlorophenol; cytotoxic to RTL-W1 rainbow trout liver cells (EC50 = 1.55 μM in a neutral red assay); increases production of ROS, inhibits apoptosis, and induces loss of the mitochondrial membrane potential and necrosis in splenocytes; induces GSH depletion in mouse liver
Catalog No. 50-299-3833 Supplier Cayman Chemical Supplier No. 2323110 G
A metabolite of pentachlorophenol; cytotoxic to RTL-W1 rainbow trout liver cells (EC50 = 1.55 μM in a neutral red assay); increases production of ROS, inhibits apoptosis, and induces loss of the mitochondrial membrane potential and necrosis in splenocytes; induces GSH depletion in mouse liver