Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachlorohydroquinone, MP Biomedicals

Catalog No. ICN218651 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN218651 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN218651 Supplier MP Biomedicals, Inc Supplier No. 0521865101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 87-87-6
Molecular Formula C6H2Cl4O2
Molecular Weight (g/mol) 247.88
InChI Key STOSPPMGXZPHKP-UHFFFAOYSA-N
Synonym tetrachlorohydroquinone, 2,3,5,6-tetrachlorohydroquinone, dihydro-p-chloranil, tetrachloro-p-benzohydroquinone, tetrachlorobenzoquinol, nordrosophilin a, 2,3,5,6-tetrachloro-1,4-benzenediol, perchlorohydroquinone, hydroquinone, tetrachloro, 1,4-benzenediol, 2,3,5,6-tetrachloro
PubChem CID 66603
ChEBI CHEBI:17042
IUPAC Name 2,3,5,6-tetrachlorobenzene-1,4-diol
SMILES C1(=C(C(=C(C(=C1Cl)Cl)O)Cl)Cl)O

Specifications

Quantity 100 g
Formula Weight 247.89
Chemical Name or Material Tetrachlorohydroquinone
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.