Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachlorophthalic Acid Hemihydrate 98.0+%, TCI America™
SDP

Catalog No. T0070500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0070500G 500 g
T007025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0070500G Supplier TCI America Supplier No. T0070500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 632-58-6
Molecular Formula C8H2Cl4O4
Molecular Weight (g/mol) 303.90
MDL Number MFCD00053309
InChI Key WZHHYIOUKQNLQM-UHFFFAOYSA-N
PubChem CID 12442
IUPAC Name tetrachlorobenzene-1,2-dicarboxylic acid
SMILES OC(=O)C1=C(Cl)C(Cl)=C(Cl)C(Cl)=C1C(O)=O

Specifications

Color White
Quantity 500 g
Formula Weight 303.90
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrachlorophthalic Acid Hemihydrate
EINECSNumber (3)-0062
RTECSNumber TI2795000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.