Learn More
YSI 3P LifeScience IDS digital conductivity cells TetraCon 925 for universal applications
Catalog No. NC1703039
Encompass_Preferred
Universal digital conductiviy measurement in aqueous samples. DS conductivity measuring cells – Digital universal measurements in different samples with proven four electrode technology. 1.5 m fixed cable with waterproof digital connector.
Catalog No. NC1703039 Supplier YSI 3P LifeScience Supplier No. 301710
Universal digital conductiviy measurement in aqueous samples. DS conductivity measuring cells – Digital universal measurements in different samples with proven four electrode technology. 1.5 m fixed cable with waterproof digital connector.