Learn More
Cole-Parmer WTW TetraCon 925-3 IDS Conductivity Probe, 2000 mS; 3 m Cable
Catalog No. NC0332977 Shop All Cole-Parmer Products
Encompass
The range of this conductivity probe is 10 S to 2000 mS at 0.5 % of value.
Catalog No. NC0332977 Supplier Cole-Parmer Supplier No. 9957538
The range of this conductivity probe is 10 S to 2000 mS at 0.5 % of value.