Learn More
AdipoGen Tetraconazole,Chemical. CAS: 112281-77-3
SDP
Catalog No. 501177756
Encompass_Preferred
Tetraconazole, AdipoGen Life Sciences. Chemical. CAS: 112281-77-3. Formula: C13H11Cl2F4N3O. MW: 372.2. Synthetic. Triazole systemic DMI-fungicide (DeMethylation Inhibitor). Mode of action is through inhibition of C14-demethylase, which is important for sterol biosynthesis (erg11/cpy51). Sterols are needed for membrane structure and function, making them essential for the development of functional cell walls. Therefore, these fungicides result in abnormal fungal growth and eventually death. Compound can be used as analytical reference material. 112281-77-3
Catalog No. 50-117-7756 Supplier AdipoGen Supplier No. CDXT0115M500
Tetraconazole, AdipoGen Life Sciences. Chemical. CAS: 112281-77-3. Formula: C13H11Cl2F4N3O. MW: 372.2. Synthetic. Triazole systemic DMI-fungicide (DeMethylation Inhibitor). Mode of action is through inhibition of C14-demethylase, which is important for sterol biosynthesis (erg11/cpy51). Sterols are needed for membrane structure and function, making them essential for the development of functional cell walls. Therefore, these fungicides result in abnormal fungal growth and eventually death. Compound can be used as analytical reference material. 112281-77-3