Learn More
FUJIFILM Biosciences Tetraconazole Ref Material CRM
Catalog No. 5000450321
Encompass_Preferred
Tetraconazole CRM is a certified reference material used in pesticide residue analysis and GC-based quality control applications Supplied as a clear liquid with 98% purity Tetraconazole Reference Material from FUJIFILM Wako is supplied as a certified single-substance CRM with high assay suitability for trace-level quantification and quality control in pesticide residue testing using GC methods Provided in sealed glass vials to maintain integrity
Catalog No. 50-004-50321 Supplier FUJIFILM Biosciences Supplier No. 20519101
Tetraconazole CRM is a certified reference material used in pesticide residue analysis and GC-based quality control applications Supplied as a clear liquid with 98% purity Tetraconazole Reference Material from FUJIFILM Wako is supplied as a certified single-substance CRM with high assay suitability for trace-level quantification and quality control in pesticide residue testing using GC methods Provided in sealed glass vials to maintain integrity