Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetracontane, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014169 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4169 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4169 Supplier MilliporeSigma Supelco Supplier No. 87086250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Featured Industries: Petroleum
TRUSTED_SUSTAINABILITY

Specifications

CAS 4181-95-7
Physical Form Neat
Linear Formula CH3(CH2)38 CH3
MDL Number MFCD00015267
Melting Point 80°C to 83°C; 80°C to 84°C
Synonym n-Tetracontane
Type Analytical Standard
Quantity 250 mg
Formula Weight 563.08
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.