Learn More
Medchemexpress LLC Tetracosactide acetate | 60189-34-6 | 99.35% | 1 MG
SDP
Catalog No. 5000426729
Encompass_Preferred
Tetracosactide acetate (Tetracosactrin acetate) is the acetate salt form of Tetracosactide, an analogue of adrenocorticotrophic hormone (ACTH). It acts as an agonist for the melanocortin-4 receptor (MC4R), activating human MC4R with an EC50 of 0.65 nM. Tetracosactide acetate stimulates the release of corticosteroids like cortisol from the adrenal gland and is used for research on various conditions.
  • Acts as an agonist for the melanocortin-4 receptor (MC4R)
  • Stimulates the release of corticosteroids such as cortisol
  • Researched for ulcerative colitis and Crohn's disease
  • Researched for juvenile/adult rheumatoid arthritis and osteoarthrosis
  • Promotes protein synthesis in adrenal slices in vitro
  • Reduces oxidative stress markers in vivo
Catalog No. 50-004-26729 Supplier Medchemexpress LLC Supplier No. HYP0060A1MG
Tetracosactide acetate (Tetracosactrin acetate) is the acetate salt form of Tetracosactide, an analogue of adrenocorticotrophic hormone (ACTH). It acts as an agonist for the melanocortin-4 receptor (MC4R), activating human MC4R with an EC50 of 0.65 nM. Tetracosactide acetate stimulates the release of corticosteroids like cortisol from the adrenal gland and is used for research on various conditions.
  • Acts as an agonist for the melanocortin-4 receptor (MC4R)
  • Stimulates the release of corticosteroids such as cortisol
  • Researched for ulcerative colitis and Crohn's disease
  • Researched for juvenile/adult rheumatoid arthritis and osteoarthrosis
  • Promotes protein synthesis in adrenal slices in vitro
  • Reduces oxidative stress markers in vivo