Learn More
Medchemexpress LLC Tetracosactide (acetate) | 60189-34-6 | 99.4% | 25 MG
SDP
Catalog No. 5000426971
Encompass_Preferred
Tetracosactide acetate is the acetate salt form of Tetracosactide. It is an analogue of adrenocorticotrophic hormone (ACTH) and an agonist for melanocortin-4 receptor (MC4R) that activates human MC4R with an EC50 of 0.65 nM. It can stimulate the release of corticosteroids such as cortisol from the adrenal gland.
  • Acetate salt form of Tetracosactide
  • Analogue of adrenocorticotrophic hormone (ACTH)
  • Agonist for melanocortin-4 receptor (MC4R)
  • Activates human MC4R with an EC50 of 0.65 nM
  • Stimulates the release of corticosteroids like cortisol
  • Used for research of ulcerative colitis and Crohn's disease, juvenile/adult rheumatoid arthritis and osteoarthrosis
Catalog No. 50-004-26971 Supplier Medchemexpress LLC Supplier No. 25MGHYP0060A
Tetracosactide acetate is the acetate salt form of Tetracosactide. It is an analogue of adrenocorticotrophic hormone (ACTH) and an agonist for melanocortin-4 receptor (MC4R) that activates human MC4R with an EC50 of 0.65 nM. It can stimulate the release of corticosteroids such as cortisol from the adrenal gland.
  • Acetate salt form of Tetracosactide
  • Analogue of adrenocorticotrophic hormone (ACTH)
  • Agonist for melanocortin-4 receptor (MC4R)
  • Activates human MC4R with an EC50 of 0.65 nM
  • Stimulates the release of corticosteroids like cortisol
  • Used for research of ulcerative colitis and Crohn's disease, juvenile/adult rheumatoid arthritis and osteoarthrosis