Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetracosane, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015053 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5053 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5053 Supplier MilliporeSigma Supelco Supplier No. 870895G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetracosane belongs to the group of saturated hydrocarbons, which is found in lubricant oils.

TRUSTED_SUSTAINABILITY

Specifications

CAS 646-31-1
Physical Form Neat
Linear Formula CH3(CH2)22 CH3
MDL Number MFCD00009352
Melting Point 49°C to 52°C; 49°C to 52°C
Boiling Point 391°C
Quantity 5 g
Formula Weight 338.65
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.