Learn More
Medchemexpress LLC Tetracosane-d50 | 16416-32-3 | 5 MG
SDP
Catalog No. 5000417554
Encompass_Preferred
Tetracosane-d50 is the deuterium labeled Tetracosane, intended for research purposes only and not for patient use. It serves as a tracer and an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes into drug molecules, is commonly used for quantitation during drug development and can influence pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-17554 Supplier Medchemexpress LLC Supplier No. HYN7933S5MG
Tetracosane-d50 is the deuterium labeled Tetracosane, intended for research purposes only and not for patient use. It serves as a tracer and an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes into drug molecules, is commonly used for quantitation during drug development and can influence pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs