Learn More
Medchemexpress LLC Tetracosane-d50 | 16416-32-3 | 50 MG
SDP
Catalog No. 5000417573
Encompass_Preferred
Tetracosane-d50 is a deuterium labeled version of Tetracosane, intended for research use only. It serves as a tracer and an internal standard for quantitative analysis using various spectroscopic techniques. The process of deuteration, incorporating stable heavy isotopes, has been noted for its potential to impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled compound
  • For research use only
Catalog No. 50-004-17573 Supplier Medchemexpress LLC Supplier No. HYN7933S50MG
Add to Cart
Add to Cart
Tetracosane-d50 is a deuterium labeled version of Tetracosane, intended for research use only. It serves as a tracer and an internal standard for quantitative analysis using various spectroscopic techniques. The process of deuteration, incorporating stable heavy isotopes, has been noted for its potential to impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled compound
  • For research use only