Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetracosanol, 99+%

Catalog No. p-3758774
Change view
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC258151000 100 mg Glass bottle
AC258150010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC258151000 Supplier Thermo Scientific Chemicals Supplier No. 258151000
Only null left
Add to Cart

CAS: 506-51-4 | C24H50O | 354.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-51-4
Molecular Formula C24H50O
Molecular Weight (g/mol) 354.66
MDL Number MFCD00042662
InChI Key TYWMIZZBOVGFOV-UHFFFAOYSA-N
Synonym 1-tetracosanol, tetracosanol, lignoceryl alcohol, lignocerol, n-tetracosanol, tetracosyl alcohol, lignoceric alcohol, unii-2n0pi37ioc, n-tetracosanol-1, 2n0pi37ioc
PubChem CID 10472
ChEBI CHEBI:77413
IUPAC Name tetracosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCCCCCO

Specifications

Melting Point 75°C to 77°C
Color White
Flash Point 218°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3(CH2)23OH
Quantity 100 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in hydrocarbons,alcohols,ether,ketones,,specially when hot
Formula Weight 354.65
Percent Purity 99+%
Physical Form Shiny Flakes or Fluffy Powder
Chemical Name or Material Tetracosanol
Show More Show Less
EINECSNumber 208-043-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.