Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetracyanoethylene 98.0+%, TCI America™
SDP

Catalog No. T007725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T007725G 25 g
T00775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T007725G Supplier TCI America Supplier No. T007725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 670-54-2
Molecular Formula C6N4
Molecular Weight (g/mol) 128.094
MDL Number MFCD00001850
InChI Key NLDYACGHTUPAQU-UHFFFAOYSA-N
Synonym tetracyanoethylene, tetracyanoethene, ethylenetetracarbonitrile, tcne, ethenetetracarbonitrile, ethene, tetracyano, 1,1,2,2-ethenetetracarbonitrile, tetrakyanethylen, 1,1,2,2-tetracyanoethene, 1,1,2,2-tetracyanoethylene
PubChem CID 12635
IUPAC Name ethene-1,1,2,2-tetracarbonitrile
SMILES C(#N)C(=C(C#N)C#N)C#N

Specifications

Melting Point 198°C
Boiling Point 125°C
Quantity 25 g
UN Number 3439
Formula Weight 128.09
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Tetracyanoethylene
RTECSNumber KM7300000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.