Learn More
Apexbio Technology LLC Tetracycline(Synonyms: Achromycin, Sumycin, Tetracyn, Tetrex, Panmycin, Robitet, Polycycline, Cyclopar), 200mg, CAS: 60-54-8.
SDP
Catalog No. 5000061184
Encompass_Preferred
Tetracycline (CAS 60-54-8) is a broad-spectrum polyketide antibiotic originally isolated from Streptomyces species It functions primarily through reversible binding to the bacterial 30S ribosomal subunit disrupting the interaction of aminoacyl-tRNA with the ribosomal acceptor site and thereby inhibiting bacterial protein synthesis Additionally tetracycline interacts partially with the 50S ribosomal subunit and may compromise bacterial membrane integrity causing leakage of intracellular components Tetracycline remains widely employed in microbiological research as an antibiotic selection marker and a tool for investigating ribosomal functions
Catalog No. 50-000-61184 Supplier Apexbio Technology LLC Supplier No. C6589200
Tetracycline (CAS 60-54-8) is a broad-spectrum polyketide antibiotic originally isolated from Streptomyces species It functions primarily through reversible binding to the bacterial 30S ribosomal subunit disrupting the interaction of aminoacyl-tRNA with the ribosomal acceptor site and thereby inhibiting bacterial protein synthesis Additionally tetracycline interacts partially with the 50S ribosomal subunit and may compromise bacterial membrane integrity causing leakage of intracellular components Tetracycline remains widely employed in microbiological research as an antibiotic selection marker and a tool for investigating ribosomal functions