Learn More
Medchemexpress LLC HY-A0107 200mg Medchemexpress, Tetracycline CAS:60-54-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA0107200MG
Encompass_Preferred
Medchemexpress, HY-A0107 200mg Tetracycline CAS:60-54-8 Tetracycline is a broad-spectrum antibiotic, exhibiting activity against a wide range of gram-positive and gram-negative bacteria. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2326