Learn More
Teknova Tetracycline, 20mg/mL in 70% Ethanol. 50mL, Sterile. 25 Pack.
SDP
Catalog No. 502746000
Encompass
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.
Catalog No. 50-274-6000 Supplier Teknova Supplier No. T219925PK
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.