Learn More
Selleck Chemical LLC Tetracycline HCl 50mg 64-75-5 NCI-c55561
SDP
Catalog No. NC1177378
Encompass_Preferred
Tetracycline HCl (NCI-c55561) is a hydrochloride salt of tetracycline that is a broad-spectrum polyketide antibiotic. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1177378 Supplier Selleck Chemical LLC Supplier No. S257450MG
Tetracycline HCl (NCI-c55561) is a hydrochloride salt of tetracycline that is a broad-spectrum polyketide antibiotic. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.