Learn More
Medchemexpress LLC Tetracycline | 60-54-8 | 98.0% | C22H24N2O8 | 100 G
SDP
Catalog No. 5000433565
Encompass_Preferred
Tetracycline is a broad-spectrum antibiotic with oral activity. It is effective against a variety of bacteria, including gram-positive and gram-negative bacteria, chlamydiae, mycoplasmas, and rickettsiae. It is used for research on infections.
  • Effective against gram-positive and gram-negative bacteria, chlamydiae, mycoplasmas, and rickettsiae.
  • Shows susceptibility of V. vulnificus with an MIC value of 0.5 μg/mL.
  • Inhibits L-amyloid aggregate formation and disassembles pre-formed fibrils.
  • Can be used in animal modeling for genetically engineered disease models.
  • Induces tumor regression based on BCR-ABL gene silencing.
  • Has been studied in numerous clinical trials for various conditions.
Catalog No. 50-004-33565 Supplier Medchemexpress LLC Supplier No. HYA0107100G
Tetracycline is a broad-spectrum antibiotic with oral activity. It is effective against a variety of bacteria, including gram-positive and gram-negative bacteria, chlamydiae, mycoplasmas, and rickettsiae. It is used for research on infections.
  • Effective against gram-positive and gram-negative bacteria, chlamydiae, mycoplasmas, and rickettsiae.
  • Shows susceptibility of V. vulnificus with an MIC value of 0.5 μg/mL.
  • Inhibits L-amyloid aggregate formation and disassembles pre-formed fibrils.
  • Can be used in animal modeling for genetically engineered disease models.
  • Induces tumor regression based on BCR-ABL gene silencing.
  • Has been studied in numerous clinical trials for various conditions.