Learn More
Medchemexpress LLC Tetracycline-d6 | 2373374-42-4 | 97.2% | C22H18D6N2O8 | 1 MG
SDP
Catalog No. 5000432189
Encompass_Preferred
Tetracycline-d6 is a deuterium-labeled variant of Tetracycline, a broad-spectrum antibiotic effective against various gram-positive and gram-negative bacteria. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-32189 Supplier Medchemexpress LLC Supplier No. HYA0107S1MG
Tetracycline-d6 is a deuterium-labeled variant of Tetracycline, a broad-spectrum antibiotic effective against various gram-positive and gram-negative bacteria. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS