Learn More
Abcam Tetracycline 10-O-ß-D-galactopyranoside, 2MG
Catalog No. 50996078
Encompass_Preferred
MW 606.6 Da. Broad spectrum polyketide antibiotic. Produced by Streptomyces genus of Actinobacteria. Acts as a protein synthesis inhibitor and is commonly used to treat acne today, and, more recently, rosacea. Historically important in reducing the number of deaths from cholera. In this galactoside analog, upon enzymatic or chemical hydrolysis of the galactoside group, the active antibiotic tetracyline is produced.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-078 Supplier Abcam Supplier No. AB2759232MG
MW 606.6 Da. Broad spectrum polyketide antibiotic. Produced by Streptomyces genus of Actinobacteria. Acts as a protein synthesis inhibitor and is commonly used to treat acne today, and, more recently, rosacea. Historically important in reducing the number of deaths from cholera. In this galactoside analog, upon enzymatic or chemical hydrolysis of the galactoside group, the active antibiotic tetracyline is produced.
The product is subject to the following: Abcam Restricted Use Statement