Learn More
U.S. Pharmacopeia Tetracycline Hydrochloride, 64-75-5, MFCD00078142, 200mg
Catalog No. NC9981238
Encompass
Molecular formula C22H24N2O8 . HCl, Molecular weight 480.9, Melting Point 417.2 - 433.4 °F (214 - 223 °C) (decomposes)
Catalog No. NC9981238 Supplier U.S. Pharmacopeia Supplier No. 1651009
Molecular formula C22H24N2O8 . HCl, Molecular weight 480.9, Melting Point 417.2 - 433.4 °F (214 - 223 °C) (decomposes)