Learn More
Zymo Research Corporation Tetracycline Hydrochloride Solution - Reagent Grade, 5x5 ml
SDP
Catalog No. 501957629
Encompass
etracycline is effective against both Gram (-) and Gram (+) bacteria. Resistance to tetracycline is conferred by the tet gene product that alters the bacterial cell membrane and transport of the antibiotic into the cell.
Catalog No. 50-195-7629 Supplier Zymo Research Corporation Supplier No. A100425
etracycline is effective against both Gram (-) and Gram (+) bacteria. Resistance to tetracycline is conferred by the tet gene product that alters the bacterial cell membrane and transport of the antibiotic into the cell.