Learn More
Zymo Research Corporation Tetracycline Hydrochloride Solution - Reagent Grade, 1x5 ml
SDP
Supplier:  Zymo Research Corporation A10045
Encompass
etracycline is effective against both Gram (-) and Gram (+) bacteria. Resistance to tetracycline is conferred by the tet gene product that alters the bacterial cell membrane and transport of the antibiotic into the cell.
Catalog No. 50-195-7630