Learn More
Research Products International Corp Tetracycline Hydrochloride, 500 Grams
SDP
Supplier:  Research Products International Corp T17000500.0
Encompass_Preferred
Tetracycline Hydrochloride is a broad-spectrum antibiotic. Tetracycline inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, preventing the attachment of aminoacyl-tRNA to the mRNA-ribosome complex. This interference disrupts the translation process and inhibits bacterial growth. 
Tetracycline HCL is effective against a broad range of bacteria, both Gram-positive and Gram-negative. It is commonly used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. It is also used in molecular biology applications, such as the selection of bacteria carrying tetracycline resistance genes in plasmids.
WARNING! This product can expose you to Tetracycline HCl, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-136-8621
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov