Learn More
Crescent Chemical Co Inc TETRACYCLINE-HCL RESEARCH GRDE
SDP
Supplier:  Crescent Chemical Co Inc SE35866.02
Encompass_Preferred
Tetracycline-HCl research grade USP.Bacteriostatic antibiotic isolated from strains of Streptomyces, active against gram positive and gram negative bacteria and also against intracellular microorganisms like Rickettsia and mycoplasma. Inhibits bacterial protein sy
Catalog No. 50-248-182