Learn More
Alkali Scientific Tetracycline Hydrochloride, 100 G
SDP
Catalog No. 5000660488
Encompass_Preferred
Tetracycline Hydrochloride, available in a 100g powder form, is a broad-spectrum antibiotic used to treat a variety of bacterial infections. It is effective against both Gram-positive and Gram-negative bacteria and works by inhibiting bacterial protein synthesis. Tetracycline is widely used in treating respiratory infections, urinary tract infections, and acne. The 100g size is ideal for large-scale clinical use or laboratory applications, particularly for research into antibiotic resistance, bacterial protein synthesis, and cell biology. This compound is also commonly used in agricultural and veterinary applications to treat infections in animals and promote growth.
Catalog No. 50-006-60488 Supplier Alkali Scientific Supplier No. TET001CP
Tetracycline Hydrochloride, available in a 100g powder form, is a broad-spectrum antibiotic used to treat a variety of bacterial infections. It is effective against both Gram-positive and Gram-negative bacteria and works by inhibiting bacterial protein synthesis. Tetracycline is widely used in treating respiratory infections, urinary tract infections, and acne. The 100g size is ideal for large-scale clinical use or laboratory applications, particularly for research into antibiotic resistance, bacterial protein synthesis, and cell biology. This compound is also commonly used in agricultural and veterinary applications to treat infections in animals and promote growth.