Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Tetracycline Hydrochloride

Catalog No. A39246
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A39246 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A39246 Supplier Gibco™ Supplier No. A39246
Get 15% Off Cell Culture Products
Use promo code CELLS15
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetracycline is a yellow, odorless, crystalline powder used as an agent to activate gene expression from an inducible expression plasmid in engineered cells expressing the tetracycline repressor protein

Tetracycline Hydrochloride is the hydrochloride salt of tetracycline
TRUSTED_SUSTAINABILITY

Specifications

Type Tetracycline Hydrochloride
Physical Form Powder
Quantity 500 mg
Shipping Condition Wet Ice
Can I use doxycycline instead of tetracycline as an inducer in the Expi293 Inducible Expression System?

Doxycycline may be used as an alternative inducing agent in the Expi293 Inducible System. It is similar to tetracycline in its mechanism of action. Doxycycline has been shown to have a longer half-life than tetracycline (48 hr vs. 24 hr, respectively). If the use of doxycycline is desired with the Expi293 Inducible System, we recommend performing a dose-response curve to determine the optimal concentration for induction.

How much tetracycline do I need to induce expression in Expi293F Inducible or Expi293F Inducible GnTI- cells?

We recommend starting with a tetracycline concentration of 1 µg/mL to induce the expression of your protein of interest. However, you can modulate the level of expression by treating cells with different amounts of tetracycline. We recommend performing a dose-response curve (0-1 µg/mL; 2-fold dilution) to determine the optimal concentration of tetracycline required to induce your protein of interest.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.