Learn More
Alkali Scientific Tetracycline Hydrochloride, 25 G
SDP
Catalog No. 5000661250
Encompass_Preferred
Tetracycline Hydrochloride, 25g, is a broad-spectrum antibiotic used in the treatment of various bacterial infections. It works by binding to bacterial ribosomes, preventing protein synthesis, and is effective against a wide range of pathogens. The 25g powder form is ideal for laboratories and clinical settings where medium-sized quantities are required. Tetracycline is commonly used to treat infections such as pneumonia, urinary tract infections, and acne. In research applications, tetracycline is employed to study bacterial resistance, protein translation, and gene expression. The 25g quantity is often used for preparing large batches of solution for repeated clinical or experimental use.
Catalog No. 50-006-61250 Supplier Alkali Scientific Supplier No. TET002CP
Tetracycline Hydrochloride, 25g, is a broad-spectrum antibiotic used in the treatment of various bacterial infections. It works by binding to bacterial ribosomes, preventing protein synthesis, and is effective against a wide range of pathogens. The 25g powder form is ideal for laboratories and clinical settings where medium-sized quantities are required. Tetracycline is commonly used to treat infections such as pneumonia, urinary tract infections, and acne. In research applications, tetracycline is employed to study bacterial resistance, protein translation, and gene expression. The 25g quantity is often used for preparing large batches of solution for repeated clinical or experimental use.