Learn More
AdipoGen Tetracycline hydrochloride
SDP
Catalog No. 502560011
Encompass_Preferred
Chemical. CAS 64-75-5. Formula C22H24N2O8 . HCl. MW 480.9. Synthetic Tetracycline is a broad-spectrum antibiotic that prevents bacterial growth by inhibiting protein synthesis. It binds to a single site in the 30S ribosomal subunit which prevents attachment of aminoacyl tRNA to the ribosomal acceptor site. It is used in cell biology as a selective agent in cell culture systems. Tetracycline is toxic to prokaryotic and eukaryotic cells and selects for cells harboring the bacterial tetR gene, which are resistant to the antibiotic.
Catalog No. 50-256-0011 Supplier AdipoGen Supplier No. CDXT0096G025
Chemical. CAS 64-75-5. Formula C22H24N2O8 . HCl. MW 480.9. Synthetic Tetracycline is a broad-spectrum antibiotic that prevents bacterial growth by inhibiting protein synthesis. It binds to a single site in the 30S ribosomal subunit which prevents attachment of aminoacyl tRNA to the ribosomal acceptor site. It is used in cell biology as a selective agent in cell culture systems. Tetracycline is toxic to prokaryotic and eukaryotic cells and selects for cells harboring the bacterial tetR gene, which are resistant to the antibiotic.