Learn More
Gold Biotechnology Inc Tetracycline Hydrochloride 25 g
SDP
Catalog No. NC2943383
Encompass_Preferred
Tetracycline is a broad-spectrum antibiotic targeting gram-negative and gram-positive bacteria. It also displays toxicity against certain eukaryotic cells. Tetracycline is used in multidrug resistance studies and in cell culture applications as a selective agent. Additionally, it promotes expression of the P450 proteins.
Catalog No. NC2943383 Supplier Gold Biotechnology Inc Supplier No. T10125
Tetracycline is a broad-spectrum antibiotic targeting gram-negative and gram-positive bacteria. It also displays toxicity against certain eukaryotic cells. Tetracycline is used in multidrug resistance studies and in cell culture applications as a selective agent. Additionally, it promotes expression of the P450 proteins.