Learn More
Medchemexpress LLC Tetracycline hydrochloride | 64-75-5 | 98.3% | 1 ML
SDP
Catalog No. 5000409440
Encompass_Preferred
Tetracycline hydrochloride is a broad-spectrum antibiotic with oral activity. It exhibits activity against a wide range of bacteria including gram-positive, gram-negative bacteria, chlamydiae, mycoplasmas, and rickettsiae. This compound is suitable for research on infections and its activity extends to inhibiting L-amyloid aggregate formation.
  • Active against gram-positive and gram-negative bacteria, chlamydiae, mycoplasmas, and rickettsiae
  • Useful for research on infections
  • Inhibits L-amyloid aggregate formation and disassembles pre-formed fibrils
  • Can be utilized in animal modeling for genetically engineered disease models
  • Capable of inducing tumor regression based on BCR-ABL gene silencing
Catalog No. 50-004-09440 Supplier Medchemexpress LLC Supplier No. HYB047410MM/1ML
Tetracycline hydrochloride is a broad-spectrum antibiotic with oral activity. It exhibits activity against a wide range of bacteria including gram-positive, gram-negative bacteria, chlamydiae, mycoplasmas, and rickettsiae. This compound is suitable for research on infections and its activity extends to inhibiting L-amyloid aggregate formation.
  • Active against gram-positive and gram-negative bacteria, chlamydiae, mycoplasmas, and rickettsiae
  • Useful for research on infections
  • Inhibits L-amyloid aggregate formation and disassembles pre-formed fibrils
  • Can be utilized in animal modeling for genetically engineered disease models
  • Capable of inducing tumor regression based on BCR-ABL gene silencing