Learn More
Medchemexpress LLC Tetracycline hydrochloride | 64-75-5 | 98.7% | 25 G
SDP
Catalog No. 5000406575
Encompass_Preferred
Tetracycline hydrochloride is a broad-spectrum antibiotic with oral activity, effective against a wide range of bacteria, including gram-positive and gram-negative bacteria, chlamydiae, mycoplasmas, and rickettsiae. It is suitable for research on infections.
  • Broad-spectrum antibiotic
  • Oral activity
  • Active against gram-positive and gram-negative bacteria
  • Inhibits L-amyloid aggregates
  • Disassembles pre-formed fibrils
Catalog No. 50-004-06575 Supplier Medchemexpress LLC Supplier No. 25GHYB0474
Tetracycline hydrochloride is a broad-spectrum antibiotic with oral activity, effective against a wide range of bacteria, including gram-positive and gram-negative bacteria, chlamydiae, mycoplasmas, and rickettsiae. It is suitable for research on infections.
  • Broad-spectrum antibiotic
  • Oral activity
  • Active against gram-positive and gram-negative bacteria
  • Inhibits L-amyloid aggregates
  • Disassembles pre-formed fibrils