Learn More
Medchemexpress LLC Tetracycline hydrochloride | 64-75-5 | 98.7% | 25 MG
SDP
Catalog No. 5000450596
Encompass_Preferred
Tetracycline hydrochloride is a broad-spectrum antibiotic with oral activity. It is effective against a wide range of bacteria including gram-positive, gram-negative bacteria, chlamydiae, mycoplasmas, and rickettsiae. This product is intended for research on infections.
  • Broad-spectrum antibiotic
  • Oral activity
  • Effective against gram-positive and gram-negative bacteria
  • Effective against chlamydiae, mycoplasmas, and rickettsiae
  • Light yellow to yellow solid appearance
  • Soluble in DMSO and water
Catalog No. 50-004-50596 Supplier Medchemexpress LLC Supplier No. 25MGHYB0474R
Tetracycline hydrochloride is a broad-spectrum antibiotic with oral activity. It is effective against a wide range of bacteria including gram-positive, gram-negative bacteria, chlamydiae, mycoplasmas, and rickettsiae. This product is intended for research on infections.
  • Broad-spectrum antibiotic
  • Oral activity
  • Effective against gram-positive and gram-negative bacteria
  • Effective against chlamydiae, mycoplasmas, and rickettsiae
  • Light yellow to yellow solid appearance
  • Soluble in DMSO and water