Learn More
Abcam Tetracycline hydrochloride (aqueous), antibiotic, 10ML
Catalog No. 502704566
Encompass_Preferred
MW 480.9 g/mol. Broad spectrum polyketide antibiotic with clinical uses in treating bacterial infections such as Rocky Mountain spotted fever, typush fever, tick fevers, Q fever, and Brill-Zinsser disease and to treat upper respiratory infections and acne. Tetracycline passively diffuses through protein channels in the cell membrane, binding to 30S ribosomes and inhibits protein synthesis by preventing access of aminoacyl tRNA to the acceptor site on the mRNA-ribosome complex. It also binds to the bacterial 50S ribosomal subunit, altering the membrane and causing intracellular components to leak from bacterial cells.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4566 Supplier Abcam Supplier No. AB14657910ML
MW 480.9 g/mol. Broad spectrum polyketide antibiotic with clinical uses in treating bacterial infections such as Rocky Mountain spotted fever, typush fever, tick fevers, Q fever, and Brill-Zinsser disease and to treat upper respiratory infections and acne. Tetracycline passively diffuses through protein channels in the cell membrane, binding to 30S ribosomes and inhibits protein synthesis by preventing access of aminoacyl tRNA to the acceptor site on the mRNA-ribosome complex. It also binds to the bacterial 50S ribosomal subunit, altering the membrane and causing intracellular components to leak from bacterial cells.
The product is subject to the following: Abcam Restricted Use Statement