Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetracycline Hydrochloride, G-Biosciences
SCI_ED

Catalog No. S139790
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139790 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139790 Supplier G-Biosciences Supplier No. RC871
Add to Cart
Add to Cart

Tetracycline is a bacteriostatic antibiotic with activity against both gram-positive and gram-negative bacteria.

Within the cell tetracycline binds reversibly to the 30S subunit of the ribosome, preventing the binding of aminoacyl tRNA and inhibiting protein synthesis and hence cell growth.

Chemical Identifiers

Molecular Weight (g/mol) 480.9 g/mol

Specifications

Quantity 500 g
Percent Purity >98%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.